3-(2-Methyl-1H-imidazol-1-yl)aniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR12259-1G
OR12259-250MG
APOSOR12259-1GEA
398
EUR
APOSOR12259-1G
APOSOR12259-250MG
3-(2-Methyl-1H-imidazol-1-yl)aniline 97%
3-(2-Methyl-1H-imidazol-1-yl)aniline
Formel:
C₁₀H₁₁N₃ Molecular Weight: 173,214 g/mol Schmelzpunkt: 119,5…121.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09702403 CAS-Nummer: 184098-19-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...