4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester

Lieferant: Apollo Scientific
Gefahr

Synonyme: N,N-Dimethyl-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propylamine, 2-{4-[3-(Dimethylamino)propoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, N,N-Dimethyl-3-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)propan-1-amine

OR12268-1G OR12268-250MG
APOSOR12268-1GEA 126 EUR
APOSOR12268-1G APOSOR12268-250MG
4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester
4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester
Formel: C₁₇H₂₈BNO₃
Molecular Weight: 305,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702395
CAS-Nummer: 627899-90-5
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR