3-Amino-5-hydroxybenzoic acid, N-FMOC protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (9H-Fluoren-9-yl)methyl (3-carboxy-5-hydroxyphenyl)carbamate, 3-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)-5-hydroxybenzoic acid

OR12369-1G OR12369-5G
APOSOR12369-1GEA 174 EUR
APOSOR12369-1G APOSOR12369-5G
3-Amino-5-hydroxybenzoic acid, N-FMOC protected 97%
3-Amino-5-hydroxybenzoic acid, N-FMOC protected
Formel: C₂₂H₁₇NO₅
Molecular Weight: 375,374 g/mol
Schmelzpunkt: 234…237 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682203
CAS-Nummer: 176442-21-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR