[3-(1-Methyl-1H-pyrazol-5-yl)phenyl]methylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(1-Methyl-1H-pyrazol-5-yl)benzylamine, 5-[3-(Aminomethyl)phenyl]-1-methyl-1H-pyrazole

OR12374-250MG OR12374-1G
APOSOR12374-250MGEA 163 EUR
APOSOR12374-250MG APOSOR12374-1G
[3-(1-Methyl-1H-pyrazol-5-yl)phenyl]methylamine 97%
[3-(1-Methyl-1H-pyrazol-5-yl)phenyl]methylamine
Formel: C₁₁H₁₃N₃
Molecular Weight: 187.241 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09702399
CAS-Nummer: 934570-45-3
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR