3-(1-Methyl-1H-pyrazol-5-yl)aniline 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(3-Aminophenyl)-1-methyl-1H-pyrazole

OR12382-1G OR12382-250MG
APOSOR12382-1GEA 493 EUR
APOSOR12382-1G APOSOR12382-250MG
3-(1-Methyl-1H-pyrazol-5-yl)aniline 95%
3-(1-Methyl-1H-pyrazol-5-yl)aniline
Formel: C₁₀H₁₁N₃
Molecular Weight: 173.214 g/mol
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09702400
CAS-Nummer: 910037-08-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR