4-(Pyrimidin-2-yl)benzylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-[4-(Aminomethyl)phenyl]pyrimidine, [4-(Pyrimidin-2-yl)phenyl]methylamine

OR12413-1G OR12413-250MG
APOSOR12413-1GEA 694 EUR
APOSOR12413-1G APOSOR12413-250MG
4-(Pyrimidin-2-yl)benzylamine 95%
4-(Pyrimidin-2-yl)benzylamine
Formel: C₁₁H₁₁N₃
Molecular Weight: 185,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09817489
CAS-Nummer: 885466-44-4
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR