2-(2-Methylphenyl)pyrrolidine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Pyrrolidin-2-yl)toluene, 2-(o-Tolyl)pyrrolidine

OR12510-1G OR12510-5G
APOSOR12510-1GEA 335 EUR
APOSOR12510-1G APOSOR12510-5G
2-(2-Methylphenyl)pyrrolidine 95%
2-(2-Methylphenyl)pyrrolidine
Formel: C₁₁H₁₅N
Molecular Weight: 161,243 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02663451
CAS-Nummer: 129540-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR