1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid 98%
Lieferant: Apollo Scientific
Synonyme:
trans-1-(tert-Butoxycarbonyl)-4-phenylpyrrolidine-3-carboxylic acid
OR12533-1G
OR12533-250MG
APOSOR12533-1GEA
364
EUR
APOSOR12533-1G
APOSOR12533-250MG
1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid 98%
1-[(tert-Butyl)oxycarbonyl]-4-phenylpyrrolidine-3-carboxylic acid
Formel:
C₁₆H₂₁NO₄ Molecular Weight: 291.342 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02089475 CAS-Nummer: 221142-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...