4-Bromo-2-(methylsulphonyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

OR12639-250MG OR12639-1G OR12639-5G
APOSOR12639-250MGEA 198 EUR
APOSOR12639-250MG APOSOR12639-1G APOSOR12639-5G
4-Bromo-2-(methylsulphonyl)pyridine 95%
4-Bromo-2-(methylsulphonyl)pyridine
Formel: C₆H₆BrNO₂S
Molecular Weight: 236.086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702650
CAS-Nummer: 1209459-93-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR