5-(Bromoacetyl)-3-phenylisoxazole 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one

OR1268-1G OR1268-250MG OR1268-5G
APOSOR1268-1GEA 72.5 EUR
APOSOR1268-1G APOSOR1268-250MG APOSOR1268-5G
5-(Bromoacetyl)-3-phenylisoxazole 95%
5-(Bromoacetyl)-3-phenylisoxazole
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Schmelzpunkt: 109…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173898
CAS-Nummer: 14731-14-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR