2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%
Lieferant: Apollo Scientific
Achtung
Synonyme: 1-(2-Amino-1-phenylethyl)pyrrolidine dihydrochloride, β-(Pyrrolidin-1-yl)phenethylamine dihydrochloride
OR12694-250MG OR12694-1G
APOSOR12694-250MGEA 509 EUR
APOSOR12694-250MG APOSOR12694-1G
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride
Formel: C₁₂H₁₈N₂·2ClH
Molecular Weight: 263,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445576
CAS-Nummer: 31788-96-2