2-[(Piperidin-4-yl)oxy]pyridine dihydrochloride 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[(Pyridin-2-yl)oxy]piperidine dihydrochloride
OR12806-1G
OR12806-250MG
OR12806-5G
APOSOR12806-1GEA
282
EUR
APOSOR12806-1G
APOSOR12806-250MG
APOSOR12806-5G
2-[(Piperidin-4-yl)oxy]pyridine dihydrochloride 98%
2-[(Piperidin-4-yl)oxy]pyridine dihydrochloride
Formel:
C₁₀H₁₄N₂O·2ClH Molecular Weight: 251.153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840120 CAS-Nummer: 313490-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...