3,5-Dimethylisoxazole-4-yl isocyanate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dimethyl-4-isocyanato-1,2-oxazole, 3,5-Dimethyl-4-isocyanatoisoxazole
OR1285-1G
OR1285-250MG
APOSOR1285-1GEA
197
EUR
APOSOR1285-1G
APOSOR1285-250MG
3,5-Dimethylisoxazole-4-yl isocyanate 97%
3,5-Dimethylisoxazole-4-yl isocyanate
May form small amount of sediment
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,124 g/mol Siedepunkt: 96…98 °C (15 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD00119910 CAS-Nummer: 131825-41-7 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...