4-Amino-5-cyano-2-(pyridin-3-yl)pyrimidine

Lieferant: Apollo Scientific
Achtung

OR1287-1G OR1287-250MG
APOSOR1287-1GEA 190 EUR
APOSOR1287-1G APOSOR1287-250MG
4-Amino-5-cyano-2-(pyridin-3-yl)pyrimidine
4-Amino-5-cyano-2-(pyridin-3-yl)pyrimidine
Formel: C₁₀H₇N₅
Molecular Weight: 197.196 g/mol
Schmelzpunkt: 275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084876
CAS-Nummer: 175205-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR