4-(4-Methylbenzyl)piperidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[(Piperidin-4-yl)methyl]toluene hydrochloride
OR12875-1G
OR12875-250MG
APOSOR12875-1GEA
325
EUR
APOSOR12875-1G
APOSOR12875-250MG
4-(4-Methylbenzyl)piperidine hydrochloride
4-(4-Methylbenzyl)piperidine hydrochloride
Formel:
C₁₃H₁₉N·ClH Molecular Weight: 225.758 g/mol Schmelzpunkt: 202…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840141 CAS-Nummer: 165110-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...