4-[(Piperidin-4-yl)methyl]pyridine dihydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(Pyridin-4-yl)methyl]piperidine dihydrochloride

OR12880-250MG OR12880-500MG
APOSOR12880-250MGEA 209 EUR
APOSOR12880-250MG APOSOR12880-500MG
4-[(Piperidin-4-yl)methyl]pyridine dihydrochloride
4-[(Piperidin-4-yl)methyl]pyridine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840136
CAS-Nummer: 1172465-66-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR