4-(Phenylsulphonyl)piperidine hydrochloride 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Phenyl piperidin-4-yl sulphone hydrochloride, 4-Benzenesulphonylpiperidine Hydrochloride

OR12886-1G OR12886-5G
APOSOR12886-1GEA 256 EUR
APOSOR12886-1G APOSOR12886-5G
4-(Phenylsulphonyl)piperidine hydrochloride 95+%
4-(Phenylsulphonyl)piperidine hydrochloride
Formel: C₁₁H₁₅NO₂S·ClH
Molecular Weight: 261.768 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840151
CAS-Nummer: 1172500-91-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR