2-(4-Methylphenoxy)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(p-Tolyloxy)benzoic acid, 4-(2-Carboxyphenoxy)toluene

OR12947-100MG
APOSOR12947-100MGEA 50.9 EUR
APOSOR12947-100MG
2-(4-Methylphenoxy)benzoic acid 95%
2-(4-Methylphenoxy)benzoic acid
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00439673
CAS-Nummer: 21905-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR