4-[(Piperidin-3-yl)oxy]pyridine dihydrochloride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-[(Pyridin-4-yl)oxy]piperidine dihydrochloride
OR13003-1G
OR13003-250MG
APOSOR13003-1GEA
866
EUR
APOSOR13003-1G
APOSOR13003-250MG
4-[(Piperidin-3-yl)oxy]pyridine dihydrochloride 95%
4-[(Piperidin-3-yl)oxy]pyridine dihydrochloride
Formel:
C₁₀H₁₄N₂O·2ClH Molecular Weight: 251.153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117748 CAS-Nummer: 1171481-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...