1,3,5-Tri(phenylethynyl)benzene
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,3,5-Tris(phenylethynyl)benzene
OR13103-1G
OR13103-250MG
OR13103-5G
APOSOR13103-1GEA
132
EUR
APOSOR13103-1G
APOSOR13103-250MG
APOSOR13103-5G
1,3,5-Tri(phenylethynyl)benzene
1,3,5-Tri(phenylethynyl)benzene
Formel:
C₃₀H₁₈ Molecular Weight: 378,464 g/mol Schmelzpunkt: 141…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861404 CAS-Nummer: 118688-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...