8-Hydroxy-6-nitroquinoline

Lieferant: Apollo Scientific
Achtung

OR13235-100MG OR13235-250MG
APOSOR13235-100MGEA 0 EUR
APOSOR13235-100MG APOSOR13235-250MG
8-Hydroxy-6-nitroquinoline
8-Hydroxy-6-nitroquinoline
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00466084
CAS-Nummer: 16727-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR