1-(Chloroacetyl)piperidine-4-carboxamide

Lieferant: Apollo Scientific
Achtung

OR13275-100MG OR13275-50MG
APOSOR13275-100MGEA 314 EUR
APOSOR13275-100MG APOSOR13275-50MG
1-(Chloroacetyl)piperidine-4-carboxamide
1-(Chloroacetyl)piperidine-4-carboxamide
Formel: C₈H₁₃ClN₂O₂
Molecular Weight: 204,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02237589
CAS-Nummer: 375359-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR