1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(1,2,3,4-Tetrahydroisoquinolin-1-yl)methylamine dihydrochloride
OR13424-100MG
OR13424-1G
OR13424-250MG
APOSOR13424-100MGEA
120
EUR
APOSOR13424-100MG
APOSOR13424-1G
APOSOR13424-250MG
1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
1-(Aminomethyl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride
Formel:
C₁₀H₁₄N₂·2ClH Molecular Weight: 235,153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00227022 CAS-Nummer: 140845-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...