5-Phenylthiophene-2-sulphonyl chloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(Chlorosulphonyl)-5-phenylthiophene
OR1361-1G
OR1361-250MG
APOSOR1361-1GEA
429
EUR
APOSOR1361-1G
APOSOR1361-250MG
5-Phenylthiophene-2-sulphonyl chloride 95%
5-Phenylthiophene-2-sulphonyl chloride
Formel:
C₁₀H₇ClO₂S₂ Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797460 CAS-Nummer: 97272-02-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...