2,2,4-Trimethyl-1,4,2-oxazasilinane 99%

Lieferant: Apollo Scientific
Gefahr

OR13695-1G OR13695-5G
APOSOR13695-1GEA 115 EUR
APOSOR13695-1G APOSOR13695-5G
2,2,4-Trimethyl-1,4,2-oxazasilinane 99%
2,2,4-Trimethyl-1,4,2-oxazasilinane
Formel: C₆H₁₅NOSi
Molecular Weight: 145,275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263699
CAS-Nummer: 10196-49-3
UN: 2924
ADR: 3+8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR