2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Amino-3-hydroxy-3-(4-methoxyphenyl)propionic acid, 4-(2-Amino-2-carboxy-1-hydroxyethyl)anisole, β-Hydroxy-O-methyl-DL-tyrosine
OR13802-250MG
APOSOR13802-250MGEA
232
EUR
APOSOR13802-250MG
2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid
2-Amino-3-hydroxy-3-(4-methoxyphenyl)propanoic acid
Formel:
C₁₀H₁₃NO₄ Molecular Weight: 211,215 g/mol Schmelzpunkt: 176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09998363 CAS-Nummer: 50897-30-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...