3-Piperidin-1-ylaniline 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR1384-1G
OR1384-5G
OR1384-10G
APOSOR1384-1GEA
111
EUR
APOSOR1384-1G
APOSOR1384-5G
APOSOR1384-10G
3-Piperidin-1-ylaniline 98%
3-Piperidin-1-ylaniline
Formel:
C₁₁H₁₆N₂ Molecular Weight: 176,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234760 CAS-Nummer: 27969-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...