1,4-Diazabicyclo[4.3.0]nonane

Lieferant: Apollo Scientific
Achtung

Synonyme: Octahydropyrrolo[1,2-a]pyrazine

OR13983-1G OR13983-250MG OR13983-5G
APOSOR13983-1GEA 109 EUR
APOSOR13983-1G APOSOR13983-250MG APOSOR13983-5G
1,4-Diazabicyclo[4.3.0]nonane
1,4-Diazabicyclo[4.3.0]nonane
Formel: C₇H₁₄N₂
Molecular Weight: 126,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082600
CAS-Nummer: 5654-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR