5-(Bromoacetyl)-2-oxindole 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(2-Bromoethanoyl)-2-oxindole, 5-(Bromoacetyl)indolin-2-one, 5-(Bromoacetyl)-1,3-dihydro-2H-indol-2-one

OR1417-1G OR1417-250MG
APOSOR1417-1GEA 314 EUR
APOSOR1417-1G APOSOR1417-250MG
5-(Bromoacetyl)-2-oxindole 97%
5-(Bromoacetyl)-2-oxindole
Formel: C₁₀H₈BrNO₂
Molecular Weight: 254.08 g/mol
Schmelzpunkt: >225 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08690294
CAS-Nummer: 105316-98-1
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR