5-(Bromoacetyl)-2,3-dihydro-1,4-benzodioxine

Lieferant: Apollo Scientific

Synonyme: 5-(Bromoacetyl)-1,4-benzodioxane, 2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-5-yl)ethan-1-one

OR14245-1G OR14245-250MG
APOSOR14245-1GEA 335 EUR
APOSOR14245-1G APOSOR14245-250MG
5-(Bromoacetyl)-2,3-dihydro-1,4-benzodioxine
5-(Bromoacetyl)-2,3-dihydro-1,4-benzodioxine
Formel: C₁₀H₉BrO₃
Molecular Weight: 257.081 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04972617
CAS-Nummer: 19815-97-5
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR