1,3-Benzothiazole-6-sulphonyl chloride 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Benzo[d]thiazole-6-sulphonyl chloride, 6-(Chlorosulphonyl)-1,3-benzothiazole
OR14303-1G
OR14303-250MG
APOSOR14303-1GEA
58
EUR
APOSOR14303-1G
APOSOR14303-250MG
1,3-Benzothiazole-6-sulphonyl chloride 95%
1,3-Benzothiazole-6-sulphonyl chloride
Formel:
C₇H₄ClNO₂S₂ Schmelzpunkt: 118…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086100 CAS-Nummer: 181124-40-3 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...