5-(Aminomethyl)-1-methyl-1H-imidazole 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(1-Methyl-1H-imidazol-5-yl)methylamine, 1-Methyl-5-aminomethylimidazole
OR14306-1G
OR14306-250MG
APOSOR14306-1GEA
395
EUR
APOSOR14306-1G
APOSOR14306-250MG
5-(Aminomethyl)-1-methyl-1H-imidazole 97%
5-(Aminomethyl)-1-methyl-1H-imidazole
Formel:
C₅H₉N₃ Molecular Weight: 111,145 g/mol Siedepunkt: 100…104 °C (0,3 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200858 CAS-Nummer: 486414-86-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...