3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid
OR14356-1G
OR14356-250MG
APOSOR14356-1GEA
325
EUR
APOSOR14356-1G
APOSOR14356-250MG
3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid 97%
3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid
Formel:
C₉H₈N₂O₄ Molecular Weight: 208.171 g/mol Schmelzpunkt: 167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01880302 CAS-Nummer: 849066-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...