(2R)-2-Amino-3-(pyridin-4-yl)propanoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: D-4-Pyridylalanine (R)-2-Amino-3-(4-pyridyl)propionic acid, 3-(Pyridin-4-yl)-D-alanine, 4-[(2R)-2-Amino-2-carboxyethyl]pyridine, (R)-2-Amino-3-(pyridin-4-yl)propanoic acid, (2R)-2-Amino-3-(pyridin-4-yl)propionic acid, 3-(4-Pyridyl)-D-alanine

OR14726-1G OR14726-5G
APOSOR14726-1GEA 112 EUR
APOSOR14726-1G APOSOR14726-5G
(2R)-2-Amino-3-(pyridin-4-yl)propanoic acid 95%
(2R)-2-Amino-3-(pyridin-4-yl)propanoic acid
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,177 g/mol
Schmelzpunkt: 261,3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01860874
CAS-Nummer: 37535-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR