3-Methylfur-2-yl isocyanate 90%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Isocyanato-3-methylfuran

OR14918-100MG OR14918-1G OR14918-500MG
APOSOR14918-100MGEA 364 EUR
APOSOR14918-100MG APOSOR14918-1G APOSOR14918-500MG
3-Methylfur-2-yl isocyanate 90%
3-Methylfur-2-yl isocyanate

May form small amount of sediment

Formel: C₆H₅NO₂
Molecular Weight: 123.109 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09879906
CAS-Nummer: 921938-67-2
UN: 2206
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR