Tetrahydro-2H-thiopyran-4-carboxaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyltetrahydro-2H-thiopyran
OR14931-1G
OR14931-5G
APOSOR14931-1GEA
183
EUR
APOSOR14931-1G
APOSOR14931-5G
Tetrahydro-2H-thiopyran-4-carboxaldehyde 95%
Tetrahydro-2H-thiopyran-4-carboxaldehyde
Formel:
C₆H₁₀OS Molecular Weight: 130,208 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03425266 CAS-Nummer: 50675-19-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...