6-Phenylpyridazin-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-6-phenylpyridazine, 3-Amino-6-phenyl-1,2-diazine

OR14968-250MG OR14968-100MG OR14968-1G
APOSOR14968-250MGEA 84.1 EUR
APOSOR14968-250MG APOSOR14968-100MG APOSOR14968-1G
6-Phenylpyridazin-3-amine
6-Phenylpyridazin-3-amine
Formel: C₁₀H₉N₃
Molecular Weight: 171.199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234636
CAS-Nummer: 14966-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR