3-Amino-6-(4-methoxyphenyl)pyridazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(4-Methoxyphenyl)pyridazin-3-amine

OR14971-100MG
APOSOR14971-100MGEA 21.9 EUR
APOSOR14971-100MG
3-Amino-6-(4-methoxyphenyl)pyridazine
3-Amino-6-(4-methoxyphenyl)pyridazine
Formel: C₁₁H₁₁N₃O
Molecular Weight: 201,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801353
CAS-Nummer: 4776-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR