5-(Benzylamino)-7-chloro-4-nitro-2,1,3-benzoxadiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Benzylamino)-7-chloro-4-nitrobenzofurazan, N-Benzyl-7-chloro-4-nitro-2,1,3-benzoxadiazol-5-amine

OR1529-100MG OR1529-250MG OR1529-500MG
APOSOR1529-100MGEA 87 EUR
APOSOR1529-100MG APOSOR1529-250MG APOSOR1529-500MG
5-(Benzylamino)-7-chloro-4-nitro-2,1,3-benzoxadiazole
5-(Benzylamino)-7-chloro-4-nitro-2,1,3-benzoxadiazole
Formel: C₁₃H₉ClN₄O₃
Molecular Weight: 304,689 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00576759
CAS-Nummer: 306934-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR