1-Methyl-1H-indole-6-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(1-Methyl-1H-indol-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR15740-100MG OR15740-1G OR15740-250MG
APOSOR15740-100MGEA 24.8 EUR
APOSOR15740-100MG APOSOR15740-1G APOSOR15740-250MG
1-Methyl-1H-indole-6-boronic acid, pinacol ester
1-Methyl-1H-indole-6-boronic acid, pinacol ester
Formel: C₁₅H₂₀BNO₂
Molecular Weight: 257,136 g/mol
Schmelzpunkt: 97…99,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690255
CAS-Nummer: 884507-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR