Methyl 1-phenylcyclobutane-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(Methoxycarbonyl)-1-phenylcyclobutane
OR15757-1G
OR15757-500MG
OR15757-5G
APOSOR15757-1GEA
154
EUR
APOSOR15757-1G
APOSOR15757-500MG
APOSOR15757-5G
Methyl 1-phenylcyclobutane-1-carboxylate
Methyl 1-phenylcyclobutane-1-carboxylate
Formel:
C₁₂H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11035753 CAS-Nummer: 58469-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...