3,4-Dihydro-6-(hydroxymethyl)-2H-1,5-benzodioxepine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,4-Dihydro-2H-benzo[b][1,4]dioxepin-6-ylmethanol
OR15762-1G
OR15762-5G
APOSOR15762-1GEA
223
EUR
APOSOR15762-1G
APOSOR15762-5G
3,4-Dihydro-6-(hydroxymethyl)-2H-1,5-benzodioxepine
3,4-Dihydro-6-(hydroxymethyl)-2H-1,5-benzodioxepine
Formel:
C₁₀H₁₂O₃ Molecular Weight: 180,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972614 CAS-Nummer: 499770-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...