3-Phenyl-D-phenylalaninol, N-BOC protected 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-Amino-3,3-diphenylpropan-1-ol, N-BOC protected

OR15920-1G OR15920-250MG
APOSOR15920-1GEA 122 EUR
APOSOR15920-1G APOSOR15920-250MG
3-Phenyl-D-phenylalaninol, N-BOC protected 98%
3-Phenyl-D-phenylalaninol, N-BOC protected
Formel: C₂₀H₂₅NO₃
Molecular Weight: 327,417 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05865075
CAS-Nummer: 155836-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR