2-(Pyridin-4-yl)pyrimidine-5-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR15956-1G OR15956-250MG
APOSOR15956-1GEA 0 EUR
APOSOR15956-1G APOSOR15956-250MG
2-(Pyridin-4-yl)pyrimidine-5-carboxylic acid 95%
2-(Pyridin-4-yl)pyrimidine-5-carboxylic acid
Formel: C₁₀H₇N₃O₂
Molecular Weight: 201,181 g/mol
Schmelzpunkt: 403,5…404.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09863218
CAS-Nummer: 259807-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR