3-(5-Bromothien-2-yl)pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-5-(pyridin-3-yl)thiophene
OR15965-1G
OR15965-250MG
OR15965-100MG
APOSOR15965-1GEA
270
EUR
APOSOR15965-1G
APOSOR15965-250MG
APOSOR15965-100MG
3-(5-Bromothien-2-yl)pyridine 97%
3-(5-Bromothien-2-yl)pyridine
Formel:
C₉H₆BrNS Molecular Weight: 240,12 g/mol Schmelzpunkt: 52…54,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971290 CAS-Nummer: 169050-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...