(S)-1-Methyl-3-(hydroxymethyl)piperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-(1-Methylpiperidin-3-yl)methanol, [(S)-1-Methylpiperidin-3-yl]methanol

OR15996-1G OR15996-250MG OR15996-5G
APOSOR15996-1GEA 195 EUR
APOSOR15996-1G APOSOR15996-250MG APOSOR15996-5G
(S)-1-Methyl-3-(hydroxymethyl)piperidine
(S)-1-Methyl-3-(hydroxymethyl)piperidine
Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035736
CAS-Nummer: 205194-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR