1-Benzyl-2-methylpiperazine 98%

Lieferant: Apollo Scientific

OR16021-1G OR16021-5G
APOSOR16021-1GEA 161 EUR
APOSOR16021-1G APOSOR16021-5G
1-Benzyl-2-methylpiperazine 98%
1-Benzyl-2-methylpiperazine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367737
CAS-Nummer: 29906-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR