6-Bromochroman
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-3,4-dihydro-2H-chromene
OR16541-1G
OR16541-500MG
OR16541-5G
APOSOR16541-1GEA
29
EUR
APOSOR16541-1G
APOSOR16541-500MG
APOSOR16541-5G
6-Bromochroman
6-Bromochroman
Formel:
C₉H₉BrO Molecular Weight: 213,071 g/mol Siedepunkt: 82…84 °C (0,05 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD10698725 CAS-Nummer: 3875-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...