3-Bromo-2-methylbenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2-methyl-benzenemethanol, (3-Bromo-o-tolyl)methanol, (3-Bromo-2-methylphenyl)methanol

OR16662-1G OR16662-5G OR16662-25G
APOSOR16662-1GEA 21.9 EUR
APOSOR16662-1G APOSOR16662-5G APOSOR16662-25G
3-Bromo-2-methylbenzyl alcohol 97%
3-Bromo-2-methylbenzyl alcohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
Schmelzpunkt: 101…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847531
CAS-Nummer: 83647-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR