3-Bromo-2-methylbenzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Bromo-3-(bromomethyl)-2-methylbenzene, α,3-Dibromo-o-xylene

OR16663-1G OR16663-250MG
APOSOR16663-1GEA 230 EUR
APOSOR16663-1G APOSOR16663-250MG
3-Bromo-2-methylbenzyl bromide 97%
3-Bromo-2-methylbenzyl bromide
Formel: C₈H₈Br₂
Molecular Weight: 263,957 g/mol
Schmelzpunkt: 31…32 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11847532
CAS-Nummer: 112299-62-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR